siRNA Details
| Gene Symbol: Cyp4b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016999.2 | NP_058695.2 | 4 | 383 | ||
| XM_008763976.1 | XP_008762198.1 | 4 | 457 | |||
| GenBank mRNA | BC074012.1 | - | 4 | 393 | 7100197 | |
| M29853.1 | - | 4 | 383 | |||

Cyp4b1
Genomic Map