siRNA Details
| Gene Symbol: Lcat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017024.2 | NP_058720.2 | 1 | 132 | ||
| GenBank mRNA | BC091155.1 | - | 1 | 132 | 7370790 | |
| FQ209624.1 | - | 1 | 147 | |||
| FQ218245.1 | - | 1 | 147 | |||
| FQ218883.1 | - | 1 | 150 | |||
| U62803.1 | - | 1 | 135 | |||
| X54096.1 | - | 1 | 146 | |||

Lcat
Genomic Map