siRNA Details
| Gene Symbol: Slc18a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013152.2 | NP_037284.2 | 3, 4 | 719 | ||
| XM_006252993.2 | XP_006253055.1 | 2, 3 | 919 | |||
| XM_008771067.1 | XP_008769289.1 | 2, 3 | 919 | |||
| GenBank mRNA | M97380.1 | - | 3, 4 | 748 | ||

Slc18a1
Genomic Map