siRNA Details
| Gene Symbol: ZNF140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300776.1 | NP_001287705.1 | 4 | 2467 | ||
| NM_001300777.1 | NP_001287706.1 | 5 | 2500 | |||
| NM_001300778.1 | NP_001287707.1 | 6 | 2682 | |||
| NM_003440.3 | NP_003431.2 | 5 | 2615 | |||
| XM_005266188.4 | XP_005266245.1 | 4 | 1886 | |||
| XM_011534833.2 | XP_011533135.1 | 5 | 1964 | |||
| XM_011534834.2 | XP_011533136.1 | 5 | 5755 | |||
| XM_011534835.2 | XP_011533137.1 | 5 | 5752 | |||
| XM_011534836.2 | XP_011533138.1 | 4 | 1780 | |||
| XM_011534837.2 | XP_011533139.1 | 4 | 1905 | |||
| XM_011534838.2 | XP_011533140.1 | 6 | 1977 | |||
| XM_011534839.2 | XP_011533141.1 | 6 | 1980 | |||
| XM_011534840.2 | XP_011533142.1 | 6 | 1945 | |||
| XM_017019924.1 | XP_016875413.1 | 6 | 1997 | |||
| XM_017019925.1 | XP_016875414.1 | 6 | 5822 | |||
| GenBank mRNA | AK122741.1 | - | 5 | 2616 | ||
| AK303240.1 | - | 4 | 1788 | |||
| BC022291.1 | - | 6 | 1874 | 4724560 | ||
| BC040561.1 | - | 5 | 1922 | 5268377 | ||
| U09368.1 | - | 5 | 1969 | |||

ZNF140
Genomic Map