siRNA Details
| Gene Symbol: ZNF141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011513562.2 | XP_011511864.1 | 3 | 3309 | ||
| XM_011513563.2 | XP_011511865.1 | 2 | 3067 | |||
| XM_017008591.1 | XP_016864080.1 | 4 | 3401 | |||
| GenBank mRNA | L15309.1 | - | 4 | 2456 | ||

ZNF141
Genomic Map