siRNA Details
| Gene Symbol: Tcf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013176.2 | NP_037308.1 | 6 | 554 | ||
| XM_006243345.1 | XP_006243407.1 | 6 | 412 | |||
| XM_006243346.1 | XP_006243408.1 | 6 | 412 | |||
| XM_006243347.1 | XP_006243409.1 | 6 | 412 | |||
| XM_006243348.1 | XP_006243410.1 | 6 | 412 | |||
| XM_006243349.1 | XP_006243411.1 | 6 | 412 | |||
| XM_006243350.1 | XP_006243412.1 | 6 | 412 | |||
| XM_006243351.1 | XP_006243413.1 | 4 | 265 | |||
| XM_008766218.1 | XP_008764440.1 | 4 | 4093 | |||
| GenBank mRNA | L09656.1 | - | 6 | 554 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_011544, NM_001253862, AK134037, X64840, AK143246, and others.

Tcf12
Genomic Map