siRNA Details
Gene Symbol: Got2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013177.2 | NP_037309.1 | 8 | 1034 | ||
GenBank mRNA | BC061792.1 | - | 8 | 982 | 5597941 | |
M12709.1 | - | 2 | 82 | |||
M18467.1 | - | 8 | 1005 |
Gene Symbol: LOC314123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773297.1 | XP_008771519.1 | 2 | 912 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
