siRNA Details
| Gene Symbol: Cpt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013200.1 | NP_037332.1 | 4 | 405 | ||
| XM_006242180.2 | XP_006242242.1 | 4 | 861 | |||
| GenBank mRNA | BC085761.1 | - | 4 | 417 | 7193798 | |
| D43623.1 | - | 3 | 392 | |||

Cpt1b
Genomic Map