siRNA Details
| Gene Symbol: Eif2ak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013223.1 | NP_037355.1 | 3, 4 | 416 | ||
| GenBank mRNA | BC081838.1 | - | 3, 4 | 449 | 7132671 | |
| L27707.1 | - | 3, 4 | 416 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif2ak1| Gene Symbol: Eif2ak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013223.1 | NP_037355.1 | 3, 4 | 416 | ||
| GenBank mRNA | BC081838.1 | - | 3, 4 | 449 | 7132671 | |
| L27707.1 | - | 3, 4 | 416 | |||