siRNA Details
| Gene Symbol: Adcyap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016989.2 | NP_058685.1 | 5 | 795 | ||
| XM_006245672.2 | XP_006245734.1 | 6 | 1491 | |||
| XM_006245673.2 | XP_006245735.1 | 6 | 1867 | |||
| XM_006245674.2 | XP_006245736.1 | 6 | 1820 | |||
| XM_008767418.1 | XP_008765640.1 | 6 | 1444 | |||
| XM_008767419.1 | XP_008765641.1 | 4 | 841 | |||

Adcyap1
Genomic Map