siRNA Details
| Gene Symbol: Gfap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017009.2 | NP_058705.2 | 1, 2 | 458 | ||
| XM_008768261.1 | XP_008766483.1 | 1, 2 | 524 | |||
| GenBank mRNA | BC088851.1 | - | 1, 2 | 458 | 7315338 | |
| EF094476.1 | - | 1, 2 | 529 | |||
| L27219.1 | - | 1, 2 | 462 | |||
| U03700.1 | - | 1, 2 | 482 | |||

Gfap
Genomic Map