siRNA Details
| Gene Symbol: Ass1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013157.3 | NP_037289.1 | 2, 3 | 196 | ||
| XM_006233911.2 | XP_006233973.1 | 4, 5 | 347 | |||
| GenBank mRNA | BC063146.1 | - | 4, 5 | 337 | 6888536 | |
| M36708.1 | - | 2, 3 | 196 | |||
| X12459.1 | - | 2, 3 | 196 | |||

Ass1
Genomic Map