siRNA Details
| Gene Symbol: Mxi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013160.2 | NP_037292.2 | 5, 6 | 580 | ||
| XM_006231604.1 | XP_006231666.1 | 5, 6 | 1139 | |||
| XM_006231605.1 | XP_006231667.1 | 5, 6 | 551 | |||
| XM_006231606.1 | XP_006231668.1 | 5, 6 | 710 | |||
| GenBank mRNA | AF003008.1 | - | 5, 6 | 533 | ||
| BC081824.1 | - | 5, 6 | 580 | 7133853 | ||

Mxi1
Genomic Map