siRNA Details
| Gene Symbol: Il2ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013163.1 | NP_037295.1 | 4 | 543 | ||
| GenBank mRNA | FQ228242.1 | - | 575 | |||
| M55049.1 | - | 4 | 543 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il2ra| Gene Symbol: Il2ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013163.1 | NP_037295.1 | 4 | 543 | ||
| GenBank mRNA | FQ228242.1 | - | 575 | |||
| M55049.1 | - | 4 | 543 | |||