siRNA Details
| Gene Symbol: Got2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013177.2 | NP_037309.1 | 10 | 1314 | ||
| GenBank mRNA | BC061792.1 | - | 10 | 1262 | 5597941 | |
| FQ210345.1 | - | 2 | 69 | |||
| M18467.1 | - | 10 | 1285 | |||
| Gene Symbol: LOC314123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773297.1 | XP_008771519.1 | 2 | 1189 | ||

Got2
Genomic Map