siRNA Details
Gene Symbol: Itgb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013180.1 | NP_037312.1 | 20 | 2493 | ||
GenBank mRNA | U60096.1 | - | 20 | 2493 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Itgb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013180.1 | NP_037312.1 | 20 | 2493 | ||
GenBank mRNA | U60096.1 | - | 20 | 2493 |