siRNA Details
| Gene Symbol: Hck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013185.3 | NP_037317.2 | 6, 7 | 732 | ||
| GenBank mRNA | BC078890.1 | - | 6, 7 | 732 | 7121407 | |
| M83666.1 | - | 6, 7 | 654 | |||
| S74141.1 | - | 6, 7 | 654 | |||
| X62345.1 | - | 6, 7 | 683 | |||

Hck
Genomic Map