siRNA Details
| Gene Symbol: Rheb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013216.1 | NP_037348.1 | 8 | 616 | ||
| GenBank mRNA | BC088857.1 | - | 8 | 670 | 7307969 | |
| FQ212847.1 | - | 8 | 638 | |||
| FQ214530.1 | - | 8 | 646 | |||
| FQ218725.1 | - | 8 | 638 | |||
| FQ225701.1 | - | 8 | 620 | |||
| FQ230203.1 | - | 8 | 620 | |||
| U08227.1 | - | 8 | 616 | |||

Rheb
Genomic Map