siRNA Details
| Gene Symbol: Ak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013218.1 | NP_037350.1 | 3 | 419 | ||
| GenBank mRNA | BC064656.1 | - | 3 | 421 | 5621289 | |
| D13062.1 | - | 3 | 419 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ak3| Gene Symbol: Ak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013218.1 | NP_037350.1 | 3 | 419 | ||
| GenBank mRNA | BC064656.1 | - | 3 | 421 | 5621289 | |
| D13062.1 | - | 3 | 419 | |||