siRNA Details
| Gene Symbol: Acly | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111095.1 | NP_001104565.1 | 4 | 427 | ||
| NM_016987.2 | NP_058683.2 | 4 | 427 | |||
| XM_006247254.2 | XP_006247316.1 | 4 | 420 | |||
| GenBank mRNA | BC100618.1 | - | 4 | 419 | 7934668 | |
| J05210.1 | - | 4 | 378 | |||

Acly
Genomic Map