siRNA Details
| Gene Symbol: Gfap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017009.2 | NP_058705.2 | 8 | 1220 | ||
| GenBank mRNA | BC088851.1 | - | 8 | 1220 | 7315338 | |
| L27219.1 | - | 8 | 1224 | |||
| U03700.1 | - | 8 | 1244 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gfap| Gene Symbol: Gfap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017009.2 | NP_058705.2 | 8 | 1220 | ||
| GenBank mRNA | BC088851.1 | - | 8 | 1220 | 7315338 | |
| L27219.1 | - | 8 | 1224 | |||
| U03700.1 | - | 8 | 1244 | |||