siRNA Details
| Gene Symbol: Pmp22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017037.1 | NP_058733.1 | 3 | 319 | ||
| XM_006246584.2 | XP_006246646.1 | 3 | 317 | |||
| XM_006246585.2 | XP_006246647.1 | 3 | 326 | |||
| GenBank mRNA | M69139.1 | - | 3 | 235 | ||
| S55427.1 | - | 3 | 235 | |||
| X62431.1 | - | 3 | 319 | |||

Pmp22
Genomic Map