siRNA Details
| Gene Symbol: Ppp2cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017040.1 | NP_058736.1 | 2 | 585 | ||
| GenBank mRNA | BC085926.1 | - | 2 | 569 | 7109733 | |
| M23591.1 | - | 2 | 585 | |||
| X14087.1 | - | 2 | 418 | |||
| X16044.1 | - | 2 | 320 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp2cb| Gene Symbol: Ppp2cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017040.1 | NP_058736.1 | 2 | 585 | ||
| GenBank mRNA | BC085926.1 | - | 2 | 569 | 7109733 | |
| M23591.1 | - | 2 | 585 | |||
| X14087.1 | - | 2 | 418 | |||
| X16044.1 | - | 2 | 320 | |||