siRNA Details
| Gene Symbol: Slc10a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017047.1 | NP_058743.1 | 2 | 559 | ||
| GenBank mRNA | BC091152.1 | - | 2 | 509 | 7370832 | |
| FQ218559.1 | - | 2 | 521 | |||
| M77479.1 | - | 2 | 559 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc10a1| Gene Symbol: Slc10a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017047.1 | NP_058743.1 | 2 | 559 | ||
| GenBank mRNA | BC091152.1 | - | 2 | 509 | 7370832 | |
| FQ218559.1 | - | 2 | 521 | |||
| M77479.1 | - | 2 | 559 | |||