siRNA Details
| Gene Symbol: Gabra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017069.3 | NP_058765.3 | 5, 6 | 796 | ||
| XM_006229511.2 | XP_006229573.1 | 5, 6 | 908 | |||
| GenBank mRNA | CB556618.1 | - | 4, 5 | 624 | ||
| X51991.1 | - | 5, 6 | 803 | |||

Gabra3
Genomic Map