siRNA Details
Gene Symbol: Cps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017072.1 | NP_058768.1 | 12 | 1208 | ||
GenBank mRNA | M11710.1 | - | 12 | 1347 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
