siRNA Details
| Gene Symbol: Acat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017075.2 | NP_058771.2 | 3 | 306 | ||
| GenBank mRNA | D00512.1 | - | 3 | 238 | ||
| D13921.1 | - | 3 | 238 | |||
| FM065829.1 | - | 3 | 306 | |||
| FM071064.1 | - | 1 | 94 | |||
| FQ217320.1 | - | 3 | 244 | |||
| FQ228108.1 | - | 3 | 293 | |||

Acat1
Genomic Map