siRNA Details
| Gene Symbol: Chrna5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017078.2 | NP_058774.2 | 3, 4 | 368 | ||
| GenBank mRNA | AY574255.1 | - | 3, 4 | 266 | ||
| J05231.1 | - | 3, 4 | 367 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chrna5| Gene Symbol: Chrna5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017078.2 | NP_058774.2 | 3, 4 | 368 | ||
| GenBank mRNA | AY574255.1 | - | 3, 4 | 266 | ||
| J05231.1 | - | 3, 4 | 367 | |||