siRNA Details
| Gene Symbol: Gdi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017088.2 | NP_058784.2 | 2 | 219 | ||
| GenBank mRNA | BC091156.1 | - | 2 | 223 | ||
| BC099763.1 | - | 2 | 219 | 7930350 | ||
| U07952.1 | - | 2 | 136 | |||
| X74402.1 | - | 2 | 125 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gdi1| Gene Symbol: Gdi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017088.2 | NP_058784.2 | 2 | 219 | ||
| GenBank mRNA | BC091156.1 | - | 2 | 223 | ||
| BC099763.1 | - | 2 | 219 | 7930350 | ||
| U07952.1 | - | 2 | 136 | |||
| X74402.1 | - | 2 | 125 | |||