siRNA Details
Gene Symbol: Tyro3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017092.1 | NP_058788.1 | 2 | 205 | ||
GenBank mRNA | D37880.1 | - | 2 | 205 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tyro3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017092.1 | NP_058788.1 | 2 | 205 | ||
GenBank mRNA | D37880.1 | - | 2 | 205 |