siRNA Details
| Gene Symbol: Ppp2ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017039.2 | NP_058735.1 | 2 | 346 | ||
| GenBank mRNA | BC070914.1 | - | 2 | 355 | 7104697 | |
| BC072531.1 | - | 2 | 346 | 7096821 | ||
| M33114.1 | - | 2 | 262 | |||
| X14159.1 | - | 2 | 262 | |||
| Gene Symbol: LOC103694903 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773894.1 | XP_008772116.1 | 841 | |||

Ppp2ca
Genomic Map