siRNA Details
| Gene Symbol: Bgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017087.1 | NP_058783.1 | 3 | 454 | ||
| GenBank mRNA | AY724513.1 | - | 3 | 457 | ||
| BC072480.1 | - | 3 | 462 | 7104041 | ||
| U17834.1 | - | 3 | 454 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bgn| Gene Symbol: Bgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017087.1 | NP_058783.1 | 3 | 454 | ||
| GenBank mRNA | AY724513.1 | - | 3 | 457 | ||
| BC072480.1 | - | 3 | 462 | 7104041 | ||
| U17834.1 | - | 3 | 454 | |||