siRNA Details
| Gene Symbol: Gucy1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017090.2 | NP_058786.2 | 2 | 289 | ||
| XM_006232518.2 | XP_006232580.1 | 3 | 881 | |||
| XM_008761107.1 | XP_008759329.1 | 1 | 912 | |||
| XM_008761108.1 | XP_008759330.1 | 2 | 655 | |||
| GenBank mRNA | AB096020.1 | - | 1 | 152 | ||
| BC085746.1 | - | 2 | 289 | 7190419 | ||
| M57405.1 | - | 2 | 608 | |||
| U60835.1 | - | 2 | 608 | |||

Gucy1a3
Genomic Map