siRNA Details
| Gene Symbol: Akt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017093.1 | NP_058789.1 | 6 | 805 | ||
| XM_008759108.1 | XP_008757330.1 | 7 | 3204 | |||
| XM_008759109.1 | XP_008757331.1 | 6 | 792 | |||
| XM_008759110.1 | XP_008757332.1 | 6 | 3069 | |||
| XM_008759111.1 | XP_008757333.1 | 6 | 915 | |||
| GenBank mRNA | D30041.1 | - | 6 | 805 | ||
| DQ198085.1 | - | 5 | 524 | |||

Akt2
Genomic Map