siRNA Details
| Gene Symbol: Capn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017116.2 | NP_058812.1 | 3 | 843 | ||
| GenBank mRNA | BC065306.1 | - | 3 | 843 | 5621252 | |
| L09120.1 | - | 3 | 529 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Capn2| Gene Symbol: Capn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017116.2 | NP_058812.1 | 3 | 843 | ||
| GenBank mRNA | BC065306.1 | - | 3 | 843 | 5621252 | |
| L09120.1 | - | 3 | 529 | |||