siRNA Details
| Gene Symbol: Capn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017117.1 | NP_058813.1 | 5 | 970 | ||
| XM_006234728.2 | XP_006234790.1 | 5 | 704 | |||
| XM_006234729.2 | XP_006234791.1 | 5 | 698 | |||
| XM_008762080.1 | XP_008760302.1 | 5 | 1602 | |||
| XM_008762081.1 | XP_008760303.1 | 5 | 1602 | |||
| GenBank mRNA | AF052540.1 | - | 5 | 622 | ||
| AF061726.2 | - | 5 | 675 | |||
| AF173834.1 | - | 5 | 615 | |||
| AF184950.1 | - | 5 | 615 | |||
| J05121.1 | - | 5 | 970 | |||
| U96367.1 | - | 5 | 671 | |||

Capn3
Genomic Map