siRNA Details
| Gene Symbol: Plcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017035.1 | NP_058731.1 | 4 | 683 | ||
| XM_006244063.2 | XP_006244125.1 | 4 | 582 | |||
| GenBank mRNA | EF089258.1 | - | 4 | 461 | ||
| M20637.1 | - | 4 | 683 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plcd1| Gene Symbol: Plcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017035.1 | NP_058731.1 | 4 | 683 | ||
| XM_006244063.2 | XP_006244125.1 | 4 | 582 | |||
| GenBank mRNA | EF089258.1 | - | 4 | 461 | ||
| M20637.1 | - | 4 | 683 | |||