siRNA Details
| Gene Symbol: Ppp2cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017040.1 | NP_058736.1 | 5, 6 | 1056 | ||
| GenBank mRNA | BC085926.1 | - | 5, 6 | 1040 | 7109733 | |
| FQ225122.1 | - | 1, 2 | 92 | |||
| FQ231171.1 | - | 1, 2 | 145 | |||
| M23591.1 | - | 5, 6 | 1056 | |||
| M58439.1 | - | 1, 2 | 138 | |||
| X14087.1 | - | 5, 6 | 889 | |||
| X16044.1 | - | 5, 6 | 791 | |||

Ppp2cb
Genomic Map