siRNA Details
| Gene Symbol: Slc10a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017047.1 | NP_058743.1 | 5, 6 | 1071 | ||
| GenBank mRNA | BC091152.1 | - | 4, 5 | 1021 | 7370832 | |
| FQ218559.1 | - | 4, 5 | 1034 | |||
| M77479.1 | - | 5, 6 | 1071 | |||

Slc10a1
Genomic Map