siRNA Details
| Gene Symbol: Slc4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017048.2 | NP_058744.1 | 3, 4 | 408 | ||
| XM_006235874.2 | XP_006235936.1 | 3, 4 | 865 | |||
| XM_006235875.2 | XP_006235937.1 | 3, 4 | 586 | |||
| XM_006235876.2 | XP_006235938.1 | 3, 4 | 648 | |||
| XM_006235877.2 | XP_006235939.1 | 2, 3 | 791 | |||
| XM_006235878.2 | XP_006235940.1 | 2, 3 | 710 | |||
| GenBank mRNA | AJ288902.1 | - | 2, 3 | 203 | ||
| J05166.1 | - | 3, 4 | 412 | |||

Slc4a2
Genomic Map