siRNA Details
| Gene Symbol: Egr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017086.1 | NP_058782.1 | 2 | 1085 | ||
| XM_006252239.2 | XP_006252301.1 | 2 | 1246 | |||
| XM_006252240.2 | XP_006252302.1 | 3 | 1412 | |||
| XM_008770818.1 | XP_008769040.1 | 2 | 1172 | |||
| GenBank mRNA | U12428.1 | - | 2 | 1085 | ||

Egr3
Genomic Map