siRNA Details
| Gene Symbol: Gucy1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017090.2 | NP_058786.2 | 3, 4 | 451 | ||
| XM_006232518.2 | XP_006232580.1 | 4, 5 | 1043 | |||
| XM_008761107.1 | XP_008759329.1 | 2, 3 | 1074 | |||
| XM_008761108.1 | XP_008759330.1 | 3, 4 | 817 | |||
| GenBank mRNA | AB096020.1 | - | 2, 3 | 314 | ||
| BC085746.1 | - | 3, 4 | 451 | 7190419 | ||
| M57405.1 | - | 3, 4 | 770 | |||
| U60835.1 | - | 3, 4 | 770 | |||

Gucy1a3
Genomic Map