siRNA Details
| Gene Symbol: Bmp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017105.1 | NP_058801.1 | 2 | 939 | ||
| GenBank mRNA | D63860.1 | - | 2 | 939 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bmp3| Gene Symbol: Bmp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017105.1 | NP_058801.1 | 2 | 939 | ||
| GenBank mRNA | D63860.1 | - | 2 | 939 | ||