siRNA Details
| Gene Symbol: Syn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017109.1 | NP_058805.1 | 1 | 449 | ||
| GenBank mRNA | AF056704.1 | - | 1 | 449 | ||
| Gene Symbol: LOC100910134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765288.1 | XP_008763510.1 | 2 | 744 | ||
| XM_008765289.1 | XP_008763511.1 | 3 | 847 | |||
| XM_008765290.1 | XP_008763512.1 | 2 | 720 | |||

Syn3
Genomic Map