siRNA Details
| Gene Symbol: Slco1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017111.1 | NP_058807.1 | 9 | 1078 | ||
| GenBank mRNA | FQ209854.1 | - | 9 | 1096 | ||
| L19031.1 | - | 9 | 1078 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slco1a1| Gene Symbol: Slco1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017111.1 | NP_058807.1 | 9 | 1078 | ||
| GenBank mRNA | FQ209854.1 | - | 9 | 1096 | ||
| L19031.1 | - | 9 | 1078 | |||