siRNA Details
| Gene Symbol: Clcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017137.1 | NP_058833.1 | 451 | |||
| XM_006248562.2 | XP_006248624.1 | 944 | ||||
| XM_006248563.2 | XP_006248625.1 | 944 | ||||
| GenBank mRNA | X64139.1 | - | 451 | |||
| Gene Symbol: LOC100912455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751081.3 | XP_003751129.1 | 3 | 944 | ||
| XM_006248554.2 | XP_006248616.1 | 3 | 944 | |||

Clcn2
Genomic Map