siRNA Details
| Gene Symbol: Adora2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017161.1 | NP_058857.1 | 1, 2 | 450 | ||
| GenBank mRNA | AF084241.1 | - | 1, 2 | 216 | ||
| BC161854.1 | - | 1, 2 | 458 | 9024632 | ||
| M91466.1 | - | 1, 2 | 450 | |||
More Information
Other Species Targeted
Set Membership: |
|

Adora2b
Genomic Map