siRNA Details
| Gene Symbol: Apcs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017170.2 | NP_058866.2 | 2 | 431 | ||
| XM_008769786.1 | XP_008768008.1 | 3 | 740 | |||
| GenBank mRNA | M83177.1 | - | 2 | 431 | ||
| X55761.1 | - | 2 | 513 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Apcs| Gene Symbol: Apcs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017170.2 | NP_058866.2 | 2 | 431 | ||
| XM_008769786.1 | XP_008768008.1 | 3 | 740 | |||
| GenBank mRNA | M83177.1 | - | 2 | 431 | ||
| X55761.1 | - | 2 | 513 | |||