siRNA Details
| Gene Symbol: Zfp36l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017172.1 | NP_058868.1 | 2 | 1009 | ||
| GenBank mRNA | X52590.1 | - | 2 | 1009 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp36l1| Gene Symbol: Zfp36l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017172.1 | NP_058868.1 | 2 | 1009 | ||
| GenBank mRNA | X52590.1 | - | 2 | 1009 | ||