siRNA Details
| Gene Symbol: Pkn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017175.2 | NP_058871.2 | 4 | 695 | ||
| XM_006255245.2 | XP_006255307.1 | 4 | 1735 | |||
| GenBank mRNA | BC061836.1 | - | 4 | 596 | 5599989 | |
| D26180.1 | - | 4 | 646 | |||
| FQ222217.1 | - | 4 | 695 | |||
| L35634.1 | - | 4 | 538 | |||

Pkn1
Genomic Map